2-Nitromesitylene 99%

Supplier: Thermo Scientific Chemicals
Warning

45307-0050
TS45307-0050EA 131.58 USD
TS45307-0050
2-Nitromesitylene 99%
2-Nitromesitylene
Formula: C₉H₁₁NO₂
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 41…45.5 °C
Flash Pt: 86 °C
Storage Temperature: Ambient
CAS Number: 603-71-4

Order Now

Specification Test Results

Appearance Cream to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 42 to 47 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR