1-Acetylpyrene 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(Pyren-1-yl)ethanone

44930-0050
TS44930-0050EA 120 USD
TS44930-0050
1-Acetylpyrene 97%
1-Acetylpyrene
Formula: C₁₈H₁₂O
MW: 244.29 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00021663
CAS Number: 3264-21-9

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 86 to 89 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR