2-Methoxy-4-nitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

44022-0050
TS44022-0050EA 126.26 USD
TS44022-0050
2-Methoxy-4-nitrobenzonitrile 98%
2-Methoxy-4-nitrobenzonitrile
Formula: C₈H₆N₂O₃
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD02093781
CAS Number: 101084-96-2

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
Melting point 178 to 182 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR