2,4,6-Trimethoxybenzylamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

43733-0050 43733-0010
TS43733-0010EA 85 USD
TS43733-0010 TS43733-0050
2,4,6-Trimethoxybenzylamine hydrochloride 98%
2,4,6-Trimethoxybenzylamine hydrochloride
Formula: C₁₀H₁₆ClNO₃
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00012855
CAS Number: 146548-59-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR