5-Fluoro-2-methylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Fluoro-2-methylbenzeneboronic acid

43065-0050
TS43065-0050EA 170.04 USD
TS43065-0050
5-Fluoro-2-methylphenylboronic acid 97%
5-Fluoro-2-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95
Melting Pt: 116…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095047
CAS Number: 163517-62-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 149 to 154 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR