2-Methyl-3-pyridineboronic acid hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Methylpyridin-3-yl)boronic acid hydrochloride2-Methylpyridine-3-boronic acid hydrochloride

43039-0010
TS43039-0010EA 290 USD
TS43039-0010
2-Methyl-3-pyridineboronic acid hydrochloride 97%
2-Methyl-3-pyridineboronic acid hydrochloride
Formula: C₆H₉BClNO₂
MW: 173.4100 g/mol
Melting Pt: 181…186 °C
Storage Temperature: Ambient
MDL Number: MFCD06659489
CAS Number: 1072952-34-1

Order Now

Specification Test Results

Appearance Off-white Crystalline solid
Infrared spectrum Conforms
HPLC ≥96.0 %
Sulfate (SO4) ≤0.1 %
Iron (Fe) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR