2-Chloro-3-fluorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-3-fluorobenzeneboronic acid(2-Chloro-3-fluorophenyl)boronic acid

43037-0010
TS43037-0010EA 207.72 USD
TS43037-0010
2-Chloro-3-fluorophenylboronic acid 97%
2-Chloro-3-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
MW: 174.37 g/mol
Melting Pt: 115.8…115.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363741
CAS Number: 871329-52-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 113 to 117 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR