3,5-Dimethoxybenzyl bromide 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Bromomethyl)-3,5-dimethoxybenzene

42734-0050
TS42734-0050EA 92.44 USD
TS42734-0050
3,5-Dimethoxybenzyl bromide 95%
3,5-Dimethoxybenzyl bromide
Formula: C₉H₁₁BrO₂
Melting Pt: 69…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321368
CAS Number: 877-88-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 68 to 72 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR