3-(tert-Butyldimethylsilyloxy)phenylboronic acid 96+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(tert-Butyldimethylsiloxy)benzeneboronic acid

39834-0050 39834-0010
TS39834-0010EA 85.72 USD
TS39834-0010 TS39834-0050
3-(tert-Butyldimethylsilyloxy)phenylboronic acid 96+%
3-(tert-Butyldimethylsilyloxy)phenylboronic acid
Formula: C₁₂H₂₁BO₃Si
Melting Pt: 86…91 °C
Storage Temperature: Ambient
MDL Number: MFCD03701505
CAS Number: 261621-12-9

Order Now

Specification Test Results

Appearance Off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR