6-Hydroxy-1-tetralone 98%

Supplier: Thermo Scientific Chemicals
Warning

39802-0050
TS39802-0050EA 72.01 USD
TS39802-0050
6-Hydroxy-1-tetralone 98%
6-Hydroxy-1-tetralone
Formula: C₁₀H₁₀O₂
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00156670
CAS Number: 3470-50-6

Order Now

Specification Test Results

Appearance Off-white to brown Crystalline powder
Infrared spectrum Conforms
Melting point 154 to 158 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR