(4-Bromo-2-fluorophenyl)acetonitrile 98+%

Warning

Synonyms: 2-(4-Bromo-2-fluorophenyl)acetonitrile4-Bromo-2-fluorobenzyl cyanide

38445-0250
TS38445-0250EA 256 USD
TS38445-0250
(4-Bromo-2-fluorophenyl)acetonitrile 98+%
(4-Bromo-2-fluorophenyl)acetonitrile
Formula: C₈H₅BrFN
MW: 214.0300 g/mol
Melting Pt: 52.8…53 °C
Storage Temperature: Ambient
MDL Number: MFCD07368653
CAS Number: 114897-91-5
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 50 to 54 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR