1-Methyl-1-phenylhydrazine 96%

Supplier: Thermo Scientific Chemicals
Warning

35479-0250 35479-0050
TS35479-0250EA 487.76 USD
TS35479-0250 TS35479-0050
1-Methyl-1-phenylhydrazine 96%
1-Methyl-1-phenylhydrazine
Formula: C₇H₁₀N₂
Boiling Pt: 75…78 °C (.4kPa)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007627
CAS Number: 618-40-6
EINECS: 210-547-9
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5810 to 1.5850 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR