Methyl-4-(hydroxymethyl)benzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

33084-0100
TS33084-0100EA 122.18 USD
TS33084-0100
Methyl-4-(hydroxymethyl)benzoate 98%
Methyl 4-(hydroxymethyl)benzoate
Formula: C₉H₁₀O₃
MW: 166.18
Melting Pt: 45…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009984
CAS Number: 6908-41-4
EINECS: 230-017-0

Order Now

Specification Test Results

Appearance White Powder or crystals
Infrared spectrum Conforms
Melting point 45 to 50 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR