Methyl pentafluorobenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

31226-0050 31226-0250
TS31226-0250EA 165.96 USD
TS31226-0250 TS31226-0050
Methyl pentafluorobenzoate 99%
Methyl pentafluorobenzoate
Formula: C₈H₃F₅O₂
Boiling Pt: 87…88 °C (20 mmHg)
Density: 1.53 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00012172
CAS Number: 36629-42-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR