4-Isopropylphenylhydrazine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

30615-0050
TS30615-0050EA 102.84 USD
TS30615-0050
4-Isopropylphenylhydrazine hydrochloride 98%
4-Isopropylphenylhydrazine hydrochloride
Formula: C₉H₁₅ClN₂
Melting Pt: 217 °C
Storage Temperature: Ambient
MDL Number: MFCD00013461
CAS Number: 118427-29-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige to brown Powder or flakes
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR