1-Benzyl-3-pyrrolidinone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(Phenylmethyl)-3-pyrrolidinone1-Benzylpyrrolidin-3-one

27103-0010 27103-0100 27103-0025
TS27103-0010EA 56.7 USD
TS27103-0010 TS27103-0100 TS27103-0025
1-Benzyl-3-pyrrolidinone 98%
1-Benzyl-3-pyrrolidinone
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Boiling Pt: 109 °C (.1 mmHg)
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005342
CAS Number: 775-16-6
EINECS: 212-274-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5360 to 1.5410 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR