5-Amino-1,2,4-thiadiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,4-Thiadiazol-5-amine

44987-0050 44987-0010
TS44987-0050EA 195 USD
TS44987-0050 TS44987-0010
5-Amino-1,2,4-thiadiazole 97%
5-Amino-1,2,4-thiadiazole
Formula: C₂H₃N₃S
Boiling Pt: 112…121 °C
Storage Temperature: Ambient
MDL Number: MFCD01102171
CAS Number: 7552-07-0

Order Now

Specification Test Results

Appearance Yellow Solid
Infrared spectrum Conforms
Melting point 112 to 115 °C
Water ≤0.5 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR