Isoquinoline-1-carbonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Isoquinolinecarbonitrile

44957-0010 44957-0050
TS44957-0050EA 170 USD
TS44957-0050 TS44957-0010
Isoquinoline-1-carbonitrile 98%
Isoquinoline-1-carbonitrile
Formula: C₁₀H₆N₂
MW: 154.17
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00134166
CAS Number: 1198-30-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 87 to 92 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR