2-(2-Pyrrolidinyl)pyridine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Pyrrolidin-2-ylpyridine
44674-0010
44674-0050
TS44674-0050EA
985
USD
TS44674-0050
TS44674-0010
2-(2-Pyrrolidinyl)pyridine 95%
2-(2-Pyrrolidinyl)pyridine
Formula:
C₉H₁₂N₂ MW: 148.21 g/mol Boiling Pt: 82 °C (.8 mmHg) Storage Temperature: Refrigerator |
MDL Number:
MFCD01691538 CAS Number: 77790-61-5 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless to light brown Viscous liquid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...