5-Bromo-3-fluoro-2-pyridine carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-3-fluoropyridine-2-carboxylic acid

44653-0050
TS44653-0050EA 354 USD
TS44653-0050
5-Bromo-3-fluoro-2-pyridine carboxylic acid 97%
5-Bromo-3-fluoro-2-pyridine carboxylic acid
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 165…170 °C
Storage Temperature: Ambient
MDL Number: MFCD07437865
CAS Number: 669066-91-5

Order Now

Specification Test Results

Appearance White to yellow to brown Powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure
Titration with NaOH ≥96.0 to ≤104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR