6-Quinoxaline​boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline

44552-0010
TS44552-0010EA 267 USD
TS44552-0010
6-Quinoxaline​boronic acid pinacol ester 97%
6-Quinoxaline​boronic acid pinacol ester
Formula: C₁₄H₁₇BN₂O₂
MW: 256.1080 g/mol
Boiling Pt: 130…145 °C (.6 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD11054040
CAS Number: 1167418-13-4

Order Now

Specification Test Results

Appearance Yellow to light brown Viscous liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR