4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid ethyl ester2-[(4-Ethoxycarbonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane4-(Ethoxycarbonylmethyl)phenylboronic acid Pinacol esterEthyl 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate

44500-0050
TS44500-0050EA 634 USD
TS44500-0050
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 97%
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester
Formula: C₁₆H₂₃BO₄
Melting Pt: 41…43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08704684
CAS Number: 859169-20-3

Order Now

Specification Test Results

Appearance White to off-white Low melting solid
Infrared spectrum Conforms
Melting point 38 to 46 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR