6-Bromo-1H-indazole 97%

Supplier: Thermo Scientific Chemicals
Warning

44459-0050
TS44459-0050EA 164 USD
TS44459-0050
6-Bromo-1H-indazole 97%
6-Bromo-1H-indazole
Formula: C₇H₅BrN₂
Melting Pt: 181…185 °C
Storage Temperature: Ambient
MDL Number: MFCD03265457
CAS Number: 79762-54-2

Order Now

Specification Test Results

Appearance Off-white to brown Solid
Infrared spectrum Conforms
Melting point 180 to 186 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR