9-Methylcarbazole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Methylcarbazole

44105-0050
TS44105-0050EA 94.3 USD
TS44105-0050
9-Methylcarbazole 99%
9-Methylcarbazole
Formula: C₁₃H₁₁N
Boiling Pt: 163…167 °C (.4Torr)
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00013431
CAS Number: 1484-12-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 87 to 92 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR