3-Formyl-2-methoxypyridine 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxy-pyridine-3-carbaldehyde2-Methoxy-3-pyridinecarboxaldehyde2-Methoxypyridine-3-carboxaldehyde2-Methoxynicotinaldehyde

43978-0050
TS43978-0050EA 265 USD
TS43978-0050
3-Formyl-2-methoxypyridine 96%
3-Formyl-2-methoxypyridine
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 200…201 °C (760 mmHg)
Melting Pt: 25 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD04115112
CAS Number: 71255-09-9

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR