3-(4-Methylphenyl)-1H-pyrazol-5-amine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Amino-3-(4-methylphenyl)pyrazole

43880-0050
TS43880-0050EA 201 USD
TS43880-0050
3-(4-Methylphenyl)-1H-pyrazol-5-amine 97%
3-(4-Methylphenyl)-1H-pyrazol-5-amine
Formula: C₁₀H₁₁N₃
Melting Pt: 152.3…153.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00084887
CAS Number: 78597-54-3

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 150 to 154 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR