(R)-3-(Aminomethyl)-1-boc-piperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-1-BOC-3-(Aminomethyl)piperidine
43642-0010
43642-0050
TS43642-0050EA
619
USD
TS43642-0050
TS43642-0010
(R)-3-(Aminomethyl)-1-boc-piperidine 97%
(R)-3-(Aminomethyl)-1-Boc-piperidine
Formula:
C₁₁H₂₂N₂O₂ MW: 214.31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03839876 CAS Number: 140645-23-4 |
Specification Test Results
Appearance | Colorless to light yellow Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Specific optical rotation | +10° to +20° (20°C, 589 nm) (c=1 in MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...