(R)-(-)-3-Hydroxytetrahydrofuran 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-Tetrahydrofuran-3-ol

43407-0010
TS43407-0010EA 127 USD
TS43407-0010
(R)-(-)-3-Hydroxytetrahydrofuran 98%
(R)-(-)-3-Hydroxytetrahydrofuran
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 181 °C
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00067101
CAS Number: 86087-24-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation -15.0° to -21.0° (20°C, 589 nm) (c=2.4, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR