2-(Trifluoromethyl)pyrazine 97%

Supplier: Thermo Scientific Chemicals
Warning

43261-0050
TS43261-0050EA 401 USD
TS43261-0050
2-(Trifluoromethyl)pyrazine 97%
2-(Trifluoromethyl)pyrazine
Formula: C₅H₃F₃N₂
Boiling Pt: 116…118 °C (760 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD06797734
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR