1H-Indazole-6-boronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Indazol-6-yl-6-boronic acid1H-Indazol-6-ylboronic acid

43102-0010
TS43102-0010EA 323 USD
TS43102-0010
1H-Indazole-6-boronic acid 97%
1H-Indazole-6-boronic acid
Formula: C₇H₇BN₂O₂
MW: 161.9500 g/mol
Melting Pt: 180…184 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972419
CAS Number: 885068-10-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 180 to 185 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR