6-Bromopyridine-2-carbonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromopicolinonitrile6-Bromo-2-pyridinecarbonitrile

42999-0010
TS42999-0010EA 66 USD
TS42999-0010
6-Bromopyridine-2-carbonitrile 98%
6-Bromopyridine-2-carbonitrile
Formula: C₆H₃BrN₂
MW: 183.0100 g/mol
Melting Pt: 98.5…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00234142
CAS Number: 122918-25-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 103 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR