(S)-(+)-1-Boc-3-hydroxypyrrolidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate

39758-0050
TS39758-0050EA 207 USD
TS39758-0050
(S)-(+)-1-Boc-3-hydroxypyrrolidine 98+%
(S)-(+)-1-Boc-3-hydroxypyrrolidine
Formula: C₉H₁₇NO₃
Melting Pt: 60…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01317839
CAS Number: 101469-92-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or low melting solid
Infrared spectrum Conforms
HPLC ≥98.0 %
Specific optical rotation +21° to +26° (20°C, 589 nm) (C=1, in Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR