1-Chloroisoquinoline 97+%

Supplier: Thermo Scientific Chemicals
Warning

39138-0050 39138-0010
TS39138-0050EA 117 USD
TS39138-0050 TS39138-0010
1-Chloroisoquinoline 97+%
1-Chloroisoquinoline
Formula: C₉H₆ClN
Melting Pt: 31…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00024134
CAS Number: 19493-44-8

Order Now

Specification Test Results

Appearance White to yellow Crystals and/or chunks or low melting solid
Infrared spectrum Conforms
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR