2,5-Dibromo-3,4-dinitrothiophene

Supplier: Thermo Scientific Chemicals
Warning

27822-0050
TS27822-0050EA 171 USD
TS27822-0050
2,5-Dibromo-3,4-dinitrothiophene
2,5-Dibromo-3,4-dinitrothiophene
Formula: C₄Br₂N₂O₄S
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00015537
CAS Number: 52431-30-8

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 135 to 140 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR