2,5-Dihydrofuran 98%

Supplier: Thermo Scientific Chemicals
Danger

24478-1000 24478-0250
TS24478-0250EA 63 USD
TS24478-0250 TS24478-1000
2,5-Dihydrofuran 98%
2,5-Dihydrofuran
Formula: C₄H₆O
Boiling Pt: 66…67 °C (760mbar)
Density: 0.92 g/cm³
Flash Pt: –016 °C
Storage Temperature: Ambient
MDL Number: MFCD00003219
CAS Number: 1708-29-8
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR