3-Thienylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Thiopheneacetic acidThiophene-3-acetic acid

20537-0500 20537-0100
TS20537-0500EA 311 USD
TS20537-0500 TS20537-0100
3-Thienylacetic acid 98%
3-Thienylacetic acid
Formula: C₆H₆O₂S
MW: 142.18 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00005473
CAS Number: 6964-21-2
EINECS: 230-166-1

Order Now

Specification Test Results

Appearance Off-white to beige Powder or flakes
Infrared spectrum Conforms
Melting point 77 to 80 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR