4,6-Dichloro-2-(methylthio)pyrimidine 98%

Supplier: Thermo Scientific Chemicals
Danger

16303-0050 16303-0250
TS16303-0250EA 213 USD
TS16303-0250 TS16303-0050
4,6-Dichloro-2-(methylthio)pyrimidine 98%
4,6-Dichloro-2-(methylthio)pyrimidine
Formula: C₅H₄Cl₂N₂S
Boiling Pt: 135…136 °C (14 mmHg)
Melting Pt: 38…42 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006086
CAS Number: 6299-25-8
EINECS: 228-577-6
UN: 3263
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow to orange Powder or crystals
Infrared spectrum Conforms
Melting point 38 to 43 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR