2-Ethyl-2-oxazoline 99+%

Supplier: Thermo Scientific Chemicals
Danger

15998-0025 15998-5000 15998-1000
TS15998-5000EA 132 USD
TS15998-5000 TS15998-1000 TS15998-0025
2-Ethyl-2-oxazoline 99+%
2-Ethyl-2-oxazoline
Formula: C₅H₉NO
Boiling Pt: 128.4 °C (760 mmHg)
Melting Pt: –62 °C
Density: 0.98 g/cm³
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00005302
CAS Number: 10431-98-8
EINECS: 233-912-4
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4390 (20°C, 589 nm)
Water ≤0.2 %
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR