Benzo[b]thiophene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Benzothiophene
, Thianaphthene
13862-1000
13862-0250
13862-0050
TS13862-0250EA
96.84
USD
TS13862-0250
TS13862-0050
TS13862-1000
Benzo[b]thiophene 97%
Benzo[b]thiophene
Formula:
C₈H₆S MW: 134.2 g/mol Boiling Pt: 221…222 °C (760 mmHg) Melting Pt: 28…32 °C Density: 1.14 g/cm³ Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005864 CAS Number: 95-15-8 EINECS: 202-395-7 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White to yellow to pink to orange Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 28 to 33 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...