3-Pyridylacetonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Pyridin-3-yl)acetonitrile3-Pyridineacetonitrile

13197-0050 13197-0010 13197-0250
TS13197-0050EA 102 USD
TS13197-0050 TS13197-0250 TS13197-0010
3-Pyridylacetonitrile 98%
3-Pyridylacetonitrile
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 105…108 °C (1.5 mmHg)
Density: 1.1 g/cm³
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006406
CAS Number: 6443-85-2
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5280 to 1.5300 (20°C, 589 nm)
GC ≥97.5 %
UV at 290 nm A: ≤0.5
Titration with NaOH ≥98.5 %
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR