1-(3-Methoxyphenyl)piperazine dihydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

12605-0250
TS12605-0250EA 207 USD
TS12605-0250
1-(3-Methoxyphenyl)piperazine dihydrochloride 98%
1-(3-Methoxyphenyl)piperazine dihydrochloride
Formula: C₁₁H₁₈Cl₂N₂O
Melting Pt: 201…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00035285
CAS Number: 6968-76-9

Order Now

Specification Test Results

Appearance White to grey to pink Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR