5,7-Dichloro-8-quinolinol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5,7-Dichloro-8-hydroxyquinoline

11343-1000 11343-0050
TS11343-1000EA 111 USD
TS11343-1000 TS11343-0050
5,7-Dichloro-8-quinolinol 98%
5,7-Dichloro-8-quinolinol
Formula: C₉H₅Cl₂NO
MW: 214.05 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 178…183 °C
MDL Number: MFCD00006786
CAS Number: 773-76-2
EINECS: 212-258-3

Order Now

Specification Test Results

Appearance White to beige to grey Powder
Infrared spectrum Conforms
Melting point 178 to 183 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR