2-Methyl-8-quinolinol ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 8-Hydroxyquinaldine8-Hydroxy-2-methylquinoline2-Methyloxine

12187-1000
TS12187-1000EA 52.8 USD
TS12187-1000
2-Methyl-8-quinolinol ≥98%
2-Methyl-8-quinolinol
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006765
CAS Number: 826-81-3
EINECS: 212-562-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 68 to 73 °C
Titration with HClO4 ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR