4-tert-Butylbenzaldehyde 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(tert-Butyl)benzaldehyde

42652-0010 42652-2500 42652-0500
TS42652-0010EA 923 USD
TS42652-0010 TS42652-0500 TS42652-2500
4-tert-Butylbenzaldehyde 95%
4-tert-Butylbenzaldehyde
Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Boiling Pt: 249 °C (760 mmHg)
Density: 0.97 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00035742
CAS Number: 939-97-9
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5290 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR