1,10-Decanediol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,10-Dihydroxy decane

18104-1000 18104-5000
TS18104-5000EA 178 USD
TS18104-5000 TS18104-1000
1,10-Decanediol 99%
1,10-Decanediol
Formula: C₁₀H₂₂O₂
MW: 174.28 g/mol
Boiling Pt: 297 °C (760 mmHg)
Melting Pt: 71…75 °C
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00004749
CAS Number: 112-47-0
EINECS: 203-975-2

Order Now

Specification Test Results

Appearance White Powder or flakes or pellets
Infrared spectrum Conforms
Melting point 72 to 74 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR