4-Methoxy-N-methylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Methyl-p-anisidine

17722-0010 17722-0100
TS17722-0100EA 206.04 USD
TS17722-0100 TS17722-0010
4-Methoxy-N-methylaniline 98%
4-Methoxy-N-methylaniline
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 135…136 °C (19 mmHg)
Melting Pt: 33…36 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008399
CAS Number: 5961-59-1
EINECS: 227-735-1

Order Now

Specification Test Results

Appearance White to grey-brown Low melting crystalline mass
Infrared spectrum Conforms
Melting point 33 to 36 °C
GC ≥97.5 %
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR