3-Bromotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

17341-1000 17341-0100
TS17341-1000EA 185 USD
TS17341-1000 TS17341-0100
3-Bromotoluene 99%
3-Bromotoluene
Formula: C₇H₇Br
MW: 171.04 g/mol
Boiling Pt: 183.7 °C (1013 hPa)
Melting Pt: –40 °C
Density: 1.41 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00000085
CAS Number: 591-17-3
EINECS: 209-702-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5510 to 1.5530 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR