2-Methyl-1-tetralone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-3,4-dihydronaphthalen-1(2H)-one

17237-0025
TS17237-0025EA 151.98 USD
TS17237-0025
2-Methyl-1-tetralone 98%
2-Methyl-1-tetralone
Formula: C₁₁H₁₂O
MW: 160.22 g/mol
Boiling Pt: 127…131 °C (12 mmHg)
Density: 1.06 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001690
CAS Number: 1590-08-5
EINECS: 216-461-8

Order Now

Specification Test Results

Appearance Clear yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5525 to 1.5545 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR