3,4-Xylidine 99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,4-Dimethylaniline4-Amino-o-xylene

15528-5000 15528-1000 15528-0050
TS15528-5000EA 156.06 USD
TS15528-5000 TS15528-1000 TS15528-0050
3,4-Xylidine 99.5%
3,4-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 49…51 °C
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007810
CAS Number: 95-64-7
EINECS: 202-437-4
UN: 3452
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Light beige to pink to brown Crystals or crystalline mass
Infrared spectrum Conforms
Melting point 49 to 51 °C
GC ≥99.0 %
Titration with HClO4 99.0 to 101.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR