2-Chlorotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Chlorotoluene

15020-0050 15020-0025 15020-0010 15020-5000
TS15020-0010EA 58 USD
TS15020-0010 TS15020-0025 TS15020-5000 TS15020-0050
2-Chlorotoluene 98%
2-Chlorotoluene
Formula: C₇H₇Cl
MW: 126.59 g/mol
Boiling Pt: 157…159 °C (760 mmHg)
Melting Pt: –36 °C
Density: 1.08 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00000562
CAS Number: 95-49-8
EINECS: 202-424-3
UN: 2238
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5235 to 1.5265 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR